| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1455620_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1440763_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1455620_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1440763_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1455620_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1440763_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700015180.mm.1 |
TC0700001775.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700015180.mm.1 |
NM_001252072.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015180.mm.1 |
NM_001252072.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015179.mm.1 |
TC0700001775.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700015179.mm.1 |
NM_001252072.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015179.mm.1 |
NM_001252072.1 |
Refseq |
| Mouse Expression Set 430 |
1440763_PM_at |
- |
- |
| Mouse Expression Set 430 |
1440763_at |
- |
- |
| Mouse Expression Set 430 |
1455620_PM_at |
- |
- |
| Mouse Expression Set 430 |
1455620_at |
- |
- |
| Murine Genome U74v2 Set |
140710_PM_at |
- |
- |
| Murine Genome U74v2 Set |
140710_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1455620_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1455620_at |
- |
- |