Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03652647_s1
          See other ZFHX2OS GE Assays ›
          Gene Symbol:
          Zfhx2os
          Gene Name:
          zinc finger homeobox 2, opposite strand
          Gene Aliases:
          Zfhx2as
          Chromosome Location:
          Chr.14: 55073091 - 55075874 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03652647_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfhx2os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004444.2 1 54 125
          GenBank mRNA AK032589.1 - 1 54 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036788 AK032589.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036788 NR_004444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385059 AK032589.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385059 NR_004444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385059 NM_001039198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385059 AB190186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385059 AK220366.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446215_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10419966 - -
          Mouse Genome 430 2.0 Array 1446215_PM_at - -
          Mouse Genome 430 2.0 Array 1446215_at - -
          Affymetrix Mouse Clariom S Array PSR1400006041.mm.1 NR_004444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006041.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006038.mm.1 NR_004444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006038.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006040.mm.1 NR_004444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006040.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006036.mm.1 NR_004444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006036.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006037.mm.1 NR_004444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006037.mm.1 TC1400000772.mm.1 MTA TC
          Mouse Expression Set 430 1433436_s_at - -
          Mouse Expression Set 430 1428003_s_at - -
          Mouse Expression Set 430 1433436_PM_s_at - -
          Mouse Expression Set 430 1428003_PM_s_at - -
          Mouse Expression Set 430 1446215_at - -
          Mouse Expression Set 430 1446215_PM_at - -
          Mouse Genome 430A 2.0 Array 1428003_s_at - -
          Mouse Genome 430A 2.0 Array 1428003_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1433436_s_at - -
          Mouse Genome 430A 2.0 Array 1433436_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1446215_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446215_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 379903 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline