Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03806661_gH
          See other OLFR987 GE Assays ›
          Gene Symbol:
          Olfr987
          Gene Name:
          olfactory receptor 987
          Gene Aliases:
          GA_x5J8B7TRSBM-251-3, GA_x5J8B7W531T-1241080-1240524, MOR203-7P
          Chromosome Location:
          Chr.2: 85330967 - 85331896 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03806661_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr987
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011785.1 NP_001011785.1 1 644 75
          GenBank mRNA BC147165.1 - 1 720 9056167 75
          BC147166.1 - 1 720 9056185 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr987

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P316368 NM_001011785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P316368 BC147165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P316368 BC147166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P316368 NM_001011785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P316368 BC147165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P316368 BC147166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P316368 NM_001011785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P316368 BC147165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P316368 BC147166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P316368 NM_001011785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P316368 BC147165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P316368 BC147166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P316368 NM_001011785.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P316368 BC147165.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P316368 BC147166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P316368 NM_001011785.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P316368 BC147165.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P316368 BC147166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316368 NM_001011785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316368 BC147165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316368 BC147166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316368 NM_001011785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316368 BC147165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316368 BC147166.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01597687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01363088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02163680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01918639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01597688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397487 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484509 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484508 - -
          Affymetrix Mouse Clariom S Array PSR0200033672.mm.1 TC0200003890.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033670.mm.1 NM_001011785.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033670.mm.1 NM_001011785.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033670.mm.1 TC0200003890.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033669.mm.1 TC0200003890.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161703 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161703 - -
          mouse-6 v1.1 (Illumina) GI_58801339-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161703 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline