| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10417384 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417365 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417414 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417340 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417424 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417473 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417362 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417339 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417386 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417385 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417425 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417413 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417360 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417463 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417397 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417363 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417475 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417383 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417385 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417426 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417426 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417465 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417258 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417235 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417315 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417461 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417264 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417373 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10412495 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417264 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417411 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417302 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417373 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417326 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417421 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417359 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417421 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1400000016.mm.1 |
NM_001177715.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000016.mm.1 |
TC1400002780.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000109.mm.1 |
NM_029288.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000109.mm.1 |
NM_029288.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000109.mm.1 |
TC1400000006.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000184.mm.1 |
NM_001029930.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000184.mm.1 |
NM_001029930.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000184.mm.1 |
TC1400002788.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000217.mm.1 |
TC1400002790.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000012.mm.1 |
NM_001177715.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000012.mm.1 |
TC1400002780.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000015.mm.1 |
NM_001177715.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000015.mm.1 |
TC1400002780.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000066.mm.1 |
NR_033117.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000066.mm.1 |
NR_033121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000066.mm.1 |
TC1400002784.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000158.mm.1 |
NR_033388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000158.mm.1 |
TC1400002787.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000157.mm.1 |
NR_033388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000157.mm.1 |
TC1400002787.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010363.mm.1 |
NM_001024712.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010363.mm.1 |
NM_001024712.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010363.mm.1 |
TC1400002849.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400011316.mm.1 |
NM_001024706.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011316.mm.1 |
NM_001024706.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011316.mm.1 |
TC1400001519.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400020742.mm.1 |
NM_001270812.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020742.mm.1 |
TC1400002843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000063.mm.1 |
NR_033117.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000063.mm.1 |
NR_033121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000063.mm.1 |
TC1400002784.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000110.mm.1 |
NM_029288.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000110.mm.1 |
NM_029288.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000110.mm.1 |
TC1400000006.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000159.mm.1 |
NR_033388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000159.mm.1 |
TC1400002787.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000216.mm.1 |
TC1400002790.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010361.mm.1 |
NM_001024712.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010361.mm.1 |
NM_001024712.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010361.mm.1 |
TC1400002849.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400011314.mm.1 |
NM_001024706.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011314.mm.1 |
NM_001024706.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011314.mm.1 |
TC1400001519.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000155.mm.1 |
NR_033388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000155.mm.1 |
TC1400002787.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010290.mm.1 |
NM_001270812.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010290.mm.1 |
TC1400002843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010300.mm.1 |
NM_001270812.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010300.mm.1 |
TC1400002843.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010348.mm.1 |
NM_001164727.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010348.mm.1 |
NM_001164727.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010348.mm.1 |
TC1400002848.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010362.mm.1 |
NM_001024712.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010362.mm.1 |
NM_001024712.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010362.mm.1 |
TC1400002849.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010367.mm.1 |
NM_001024712.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010367.mm.1 |
NM_001024712.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010367.mm.1 |
TC1400002849.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010366.mm.1 |
NM_001024712.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010366.mm.1 |
NM_001024712.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010366.mm.1 |
TC1400002849.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400020698.mm.1 |
NR_033117.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020698.mm.1 |
NR_033121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020698.mm.1 |
TC1400002784.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000013.mm.1 |
NM_001177715.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000013.mm.1 |
TC1400002780.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000014.mm.1 |
NM_001177715.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000014.mm.1 |
TC1400002780.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000061.mm.1 |
NR_033117.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000061.mm.1 |
NR_033121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000061.mm.1 |
TC1400002784.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000160.mm.1 |
NR_033388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000160.mm.1 |
TC1400002787.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400010408.mm.1 |
NM_001270842.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400010408.mm.1 |
TC1400002854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400020699.mm.1 |
NR_033388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020699.mm.1 |
TC1400002787.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000062.mm.1 |
NR_033117.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000062.mm.1 |
NR_033121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000062.mm.1 |
TC1400002784.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000163.mm.1 |
NR_033388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000163.mm.1 |
TC1400002787.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000218.mm.1 |
TC1400002790.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400020741.mm.1 |
NM_001270812.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020741.mm.1 |
TC1400002843.mm.1 |
MTA TC |