| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000007287.mm.1 |
NM_001142963.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007287.mm.1 |
NM_001142963.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007287.mm.1 |
TC1000000907.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007286.mm.1 |
NM_001142963.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007286.mm.1 |
NM_001142963.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007286.mm.1 |
TC1000000907.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007288.mm.1 |
NM_001142963.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007288.mm.1 |
NM_001142963.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007288.mm.1 |
TC1000000907.mm.1 |
MTA TC |