Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03807424_m1
          See other UBE2QL1 GE Assays ›
          Gene Symbol:
          Ube2ql1
          Gene Name:
          ubiquitin-conjugating enzyme E2Q family-like 1
          Gene Aliases:
          3110006E14Rik
          Chromosome Location:
          Chr.13: 69702832 - 69739897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03807424_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2ql1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145162.1 NP_001138634.1 1 - 2 1339 90
          GenBank mRNA BC058543.1 - 1 - 2 274 90
          BC116979.1 - 1 - 2 389 90
          BC116981.1 - 1 - 2 389 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002279.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          nucleotide binding
          ATP binding
          ligase activity
          ubiquitin conjugating enzyme activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 AK014009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 BC058543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 BC116979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 BC116981.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 NM_001145162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 AK014009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 BC058543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 BC116979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 BC116981.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343215 NM_001145162.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51715 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112493 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102584 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 AK014009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 BC058543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 BC116979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 BC116981.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 NM_001145162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 AK014009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 BC058543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 BC116979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 BC116981.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343215 NM_001145162.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112493 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07909450 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102584 - -
          Whole Mouse Genome G4122A A_51_P343215 AK014009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 BC058543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 BC116979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 BC116981.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 NM_001145162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 AK014009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 BC058543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 BC116979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 BC116981.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343215 NM_001145162.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112493 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11087176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11087174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10600500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10786815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10889739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10776452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10574311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10600502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11052395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11052391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10634789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10439250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10756330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11052405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10600490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10819217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11052404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10600488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11083401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11052396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11087213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10439249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10783611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10699341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11087171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117279 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10410461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410462 - -
          Affymetrix HT MG-430 PM Array Plate 1452779_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10410460 - -
          Mouse Genome 430 2.0 Array 1452779_at - -
          Mouse Genome 430 2.0 Array 1447477_at - -
          Mouse Genome 430 2.0 Array 1447477_PM_at - -
          Mouse Genome 430 2.0 Array 1452779_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76980_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76980_at - -
          Affymetrix Mouse Clariom S Array PSR1300014899.mm.1 NM_001145162.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014899.mm.1 NM_001145162.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014899.mm.1 TC1300002279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014900.mm.1 NM_001145162.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014900.mm.1 NM_001145162.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014900.mm.1 TC1300002279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014898.mm.1 NM_001145162.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014898.mm.1 NM_001145162.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014898.mm.1 TC1300002279.mm.1 MTA TC
          Mouse Expression Set 430 1447477_PM_at - -
          Mouse Expression Set 430 1452779_at - -
          Mouse Expression Set 430 1452779_PM_at - -
          Mouse Expression Set 430 1447477_at - -
          Murine Genome U74v2 Set 168641_at - -
          Murine Genome U74v2 Set 168640_PM_r_at - -
          Murine Genome U74v2 Set 168641_PM_at - -
          Murine Genome U74v2 Set 168640_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1447477_at - -
          Affymetrix HT Mouse Genome 430B Array 1452779_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452779_at - -
          Affymetrix HT Mouse Genome 430B Array 1447477_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 407913 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239445 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239445 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline