Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03807425_g1
          See other HBS1L GE Assays ›
          Gene Symbol:
          Hbs1l
          Gene Name:
          Hbs1-like (S. cerevisiae)
          Gene Aliases:
          2810035F15Rik, AI326327, eRFS
          Chromosome Location:
          Chr.10: 21295979 - 21368889 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03807425_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hbs1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042593.1 NP_001036058.1 1 - 2 195 72
          NM_001145209.1 NP_001138681.1 1 - 2 195 72
          NM_019702.2 NP_062676.2 1 - 2 195 72
          GenBank mRNA AK006626.1 - 1 - 2 168 72
          AK012856.1 - 1 - 2 145 72
          AK146678.1 - 1 - 2 161 72
          AK166140.1 - 1 - 2 158 72
          AK168671.1 - 1 - 2 158 72
          AK169281.1 - 1 - 2 158 72
          AK173091.1 - 1 - 2 141 72
          BC010251.1 - 1 - 2 120 3964560 72
          BC085235.1 - 1 - 2 137 5328213 72
          BU510509.1 - 1 - 2 147 72
          CJ082265.1 - 1 - 2 195 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hbs1l

          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational elongation
          biological_process
          nucleotide binding
          translation elongation factor activity
          GTPase activity
          GTP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000001175.mm.1 NM_001042593.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001175.mm.1 NM_001042593.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001175.mm.1 NM_001145209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001175.mm.1 NM_001145209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001175.mm.1 NM_019702.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001175.mm.1 NM_019702.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001175.mm.1 TC1000000165.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001174.mm.1 NM_001042593.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001174.mm.1 NM_001042593.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001174.mm.1 NM_001145209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001174.mm.1 NM_001145209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001174.mm.1 NM_019702.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001174.mm.1 NM_019702.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001174.mm.1 TC1000000165.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001172.mm.1 NM_001042593.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001172.mm.1 NM_001042593.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001172.mm.1 NM_001145209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001172.mm.1 NM_001145209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001172.mm.1 NM_019702.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001172.mm.1 NM_019702.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001172.mm.1 TC1000000165.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline