Search
Search

Ccdc58| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159421.1 | NP_001152893.1 | 3 - 4 | 683 | 94 | |
| NM_001159422.1 | NP_001152894.1 | 4 - 5 | 460 | 94 | ||
| NM_198645.2 | NP_941047.1 | 4 - 5 | 809 | 94 | ||
| GenBank mRNA | AK135450.1 | - | 4 - 5 | 389 | 94 | |
| BB686073.1 | - | 2 - 3 | 124 | 94 | ||
| BC024406.1 | - | 4 - 5 | 373 | 94 | ||
| BC024862.1 | - | 4 - 5 | 784 | 5356370 | 94 | |
| BC053690.1 | - | 4 - 5 | 361 | 94 | ||
| CJ094333.1 | - | 3 - 4 | 264 | 94 | ||
Set Membership: |
|