Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03808833_sH
          See other OLFR156 GE Assays ›
          Gene Symbol:
          Olfr156
          Gene Name:
          olfactory receptor 156
          Gene Aliases:
          MOR262-6, OR37B, Olfr37b, mOR37b
          Chromosome Location:
          Chr.4: 43820266 - 43821433 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03808833_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr156
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019474.2 NP_062347.2 1 143 98
          GenBank mRNA BC101957.1 - 1 143 40044987 98
          BC101961.1 - 1 143 40044981 98
          BC103551.1 - 1 143 40044986 98
          BC104727.1 - 1 143 40044983 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr156

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106572 BC104727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106572 BC103551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106572 NM_019474.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106572 BC101957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106572 BC101961.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108213 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106572 BC104727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106572 BC103551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106572 NM_019474.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106572 BC101957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106572 BC101961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P524953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873816 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108213 - -
          Whole Mouse Genome G4122A A_52_P524953 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106572 BC104727.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106572 BC103551.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106572 NM_019474.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106572 BC101957.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106572 BC101961.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255459 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255462 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04000541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03254571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03907183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03210657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04045177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512631 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512636 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512632 - -
          Mouse Genome 430 2.0 Array 1422337_at - -
          Mouse Genome 430 2.0 Array 1422337_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 29846_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 29846_at - -
          Affymetrix Mouse Clariom S Array PSR0400020775.mm.1 TC0400002545.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020775.mm.1 NM_019474.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020775.mm.1 NM_019474.2 Refseq
          Mouse Expression Set 430 1422337_PM_at - -
          Mouse Expression Set 430 1422337_at - -
          Murine Genome U74v2 Set 97000_s_at - -
          Murine Genome U74v2 Set 97000_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1422337_PM_at - -
          Mouse Genome 430A 2.0 Array 1422337_at - -
          Affymetrix HT Mouse Genome 430A Array 1422337_at - -
          Affymetrix HT Mouse Genome 430A Array 1422337_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4070091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2672012 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2672012 - -
          mouse-6 v1.1 (Illumina) scl24390.1.1_240-S - -
          Illumina Sentrix mouse-6.v1 bead array 4070091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24390.1.1_240-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2672012 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2672012 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline