Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03944397_s1
          See other C330006A16RIK GE Assays ›
          Gene Symbol:
          C330006A16Rik
          Gene Name:
          RIKEN cDNA C330006A16 gene
          Gene Aliases:
          AI551216
          Chromosome Location:
          Chr.2: 26136807 - 26140506 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03944397_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C330006A16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256521.1 NP_001243450.1 2 1029 58
          NM_001256522.1 NP_001243451.1 2 869 58
          GenBank mRNA AK077073.1 - 1 48 58
          AK082757.1 - 1 51 58

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          positive regulation of viral process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P139493 AK082757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P139493 BB397899.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P139493 BC020361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P139493 AI551216.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P139493 AK077073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P139493 AK082757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P139493 BB397899.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P139493 BC020361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P139493 AI551216.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P139493 AK077073.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139493 AK082757.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139493 BB397899.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139493 BC020361.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139493 AI551216.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139493 AK077073.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01603520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01865731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21076541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433618_PM_at - -
          Mouse Genome 430 2.0 Array 1433618_at - -
          Mouse Genome 430 2.0 Array 1433618_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109299_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109299_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200026445.mm.1 NM_001256521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026445.mm.1 NM_001256521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026445.mm.1 NM_001256522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026445.mm.1 NM_001256522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026445.mm.1 TC0200003177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026444.mm.1 NM_001256521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026444.mm.1 NM_001256521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026444.mm.1 NM_001256522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026444.mm.1 NM_001256522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026444.mm.1 TC0200003177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026449.mm.1 NM_001256521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026449.mm.1 NM_001256521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026449.mm.1 TC0200003177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026447.mm.1 NM_001256522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026447.mm.1 NM_001256522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026447.mm.1 TC0200003177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026448.mm.1 NM_001256522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026448.mm.1 NM_001256522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026448.mm.1 TC0200003177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026441.mm.1 NM_001256521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026441.mm.1 NM_001256521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026441.mm.1 NM_001256522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026441.mm.1 NM_001256522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026441.mm.1 TC0200003177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026446.mm.1 NM_001256521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026446.mm.1 NM_001256521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026446.mm.1 NM_001256522.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026446.mm.1 NM_001256522.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026446.mm.1 TC0200003177.mm.1 MTA TC
          Mouse Expression Set 430 1433618_PM_at - -
          Mouse Expression Set 430 1433618_at - -
          Murine Genome U74v2 Set 95604_PM_at - -
          Murine Genome U74v2 Set 95604_at - -
          Mouse Genome 430A 2.0 Array 1433618_at - -
          Mouse Genome 430A 2.0 Array 1433618_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1433618_at - -
          Affymetrix HT Mouse Genome 430A Array 1433618_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 768839 - -
          ABI Mouse Genome Survey Microarray 784280 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103610692 - -
          Illumina mouse-6 v1.0 expression beadchip 100940132 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2445541 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258071 - -
          mouse-6 v1.1 (Illumina) GI_38074579-S - -
          Illumina Sentrix mouse-6.v1 bead array 100940132 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100940132 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2527288 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline