Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949946_m1
          See other GM2912 GE Assays ›
          Gene Symbol:
          Gm2912
          Gene Name:
          predicted gene 2912
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949946_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm2912
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK148097.1 - 278 86
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY476947.1 - 1 - 2 401 Chr. 12 - 68973873 - 68987967 [+] on Build GRCm38 86
          GenBankmRNA BY741294.1 - 2 - 3 278 Chr. 12 - 68970570 - 68988333 [+] on Build GRCm38 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream DDB1 and CUL4 associated factor 5 pseudogene Gm17821 -1.3mb Anti-sense Chr. 12 - 67656742 - 67669271 on NCBI Build GRCm38
          Downstream ribosomal protein S29 Rps29 169.8kb Anti-sense Chr. 12 - 69157722 - 69159186 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632384 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134074 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07653290 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07653267 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07653290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09841358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09867112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10351303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10075112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09966281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10424044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09867116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09903695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09826201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09867111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09903685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28512352 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline