| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100016197.mm.1 |
NR_045346.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016197.mm.1 |
NR_045346.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016197.mm.1 |
TC1100001711.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100016198.mm.1 |
NR_045346.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016198.mm.1 |
NR_045346.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016198.mm.1 |
TC1100001711.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100016199.mm.1 |
NR_045346.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016199.mm.1 |
NR_045346.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016199.mm.1 |
TC1100001711.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100016200.mm.1 |
NR_045346.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016200.mm.1 |
NR_045346.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100016200.mm.1 |
TC1100001711.mm.1 |
MTA TC |
| Murine Genome U74v2 Set |
134648_PM_at |
- |
- |
| Murine Genome U74v2 Set |
134648_at |
- |
- |