| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800017848.mm.1 |
NM_001200023.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017848.mm.1 |
NM_001200023.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017848.mm.1 |
TC0800002372.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800017845.mm.1 |
NM_001200023.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017845.mm.1 |
NM_001200023.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017845.mm.1 |
TC0800002372.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800017846.mm.1 |
NM_001200023.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017846.mm.1 |
NM_001200023.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017846.mm.1 |
TC0800002372.mm.1 |
MTA TC |