Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03951982_m1
          See other BC037156 GE Assays ›
          Gene Symbol:
          BC037156
          Gene Name:
          cDNA sequence BC037156
          Gene Aliases:
          FDC-SP
          Chromosome Location:
          Chr.5: 87908779 - 87914171 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03951982_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC037156
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC037156.1 - 4 - 5 349 101
          JQ408704.1 - 4 - 5 339 101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF301466.1 - 4 - 5 338 Chr. 5 - 87908779 - 87914173 [+] on Build GRCm38 101
          GenBankmRNA BF300338.1 - 4 - 5 319 Chr. 5 - 87908798 - 87914173 [+] on Build GRCm38 101
          GenBankmRNA BF301743.1 - 4 - 5 339 Chr. 5 - 87908781 - 87914173 [+] on Build GRCm38 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700066N21 gene 1700066N21Rik Anti-sense Chr. 5 - 87908588 - 87979351 on NCBI Build GRCm38
          Downstream casein kappa Csn3 AW208918,
          CSN10,
          Csnk
          11.4kb Sense Chr. 5 - 87925575 - 87932545 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974632 BC037156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974632 BF301743.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974632 BC037156.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974632 BF301743.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04388978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870230 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10522892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522890 - -
          Affymetrix Mouse Gene 1.1 ST Array 10522889 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2494135 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0