Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952363_m1
          See other 4933403O08RIK GE Assays ›
          Gene Symbol:
          4933403O08Rik
          Gene Name:
          RIKEN cDNA 4933403O08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 112171438 - 112249350 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952363_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933403O08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177389.1 NP_001170860.1 2 - 3 987 82
          GenBank mRNA AK016644.1 - 2 - 3 984 82
          BC115851.1 - 2 - 3 539 82
          BC127170.1 - 1 - 2 875 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001122.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102562 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978601 NM_001177389.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102562 - -
          Whole Mouse Genome G4122A A_52_P375617 AK016644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375617 BC115851.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375617 NM_001177389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375617 BC127170.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125641 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14648491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15000653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14544421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15149027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14548794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32602590 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453533_PM_at - -
          Mouse Genome 430 2.0 Array 1453533_at - -
          Mouse Genome 430 2.0 Array 1453533_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71030_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71030_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00007334.mm.1 NM_001177389.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007334.mm.1 NM_001177389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007334.mm.1 TC0X00001122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007335.mm.1 NM_001177389.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007335.mm.1 NM_001177389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007335.mm.1 TC0X00001122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007332.mm.1 NM_001177389.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007332.mm.1 NM_001177389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007332.mm.1 TC0X00001122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007333.mm.1 NM_001177389.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007333.mm.1 NM_001177389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007333.mm.1 TC0X00001122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007331.mm.1 NM_001177389.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007331.mm.1 NM_001177389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007331.mm.1 TC0X00001122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007330.mm.1 NM_001177389.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007330.mm.1 NM_001177389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007330.mm.1 TC0X00001122.mm.1 MTA TC
          Mouse Expression Set 430 1453533_at - -
          Mouse Expression Set 430 1453533_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453533_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453533_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 781638 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2648155 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2648155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-z8rtm:80/100.66.72.105:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline