Search
Search

Mapk1ip1l| Gene Symbol: Mapk1ip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034789.1 | - | 1 | 3150 | 79 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppressor of cytokine signaling 4 | Socs4 |
3110032M18Rik, A730004F22Rik, AI427843, AU015727, AU015859, Socs7 |
-21.3kb | Sense | Chr. 14 - 47277143 - 47291591 on NCBI Build GRCm38 |
| Downstream |
lectin, galactose binding, soluble 3 |
Lgals3 |
GBP, L-34, Mac-2, gal3 |
57.2kb | Sense | Chr. 14 - 47373860 - 47386167 on NCBI Build GRCm38 |
Set Membership: |
|