Search
Search
| Gene Symbol: Acd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012638.1 | NP_001012656.1 | 1 - 2 | 178 | 73 | |
| GenBank mRNA | AK032683.1 | - | 1 - 2 | 158 | 73 | |
| AK052768.1 | - | 169 | 73 | |||
| AK145959.1 | - | 1 - 2 | 163 | 73 | ||
| AK160599.1 | - | 1 - 2 | 138 | 73 | ||
| AK171345.1 | - | 1 - 2 | 138 | 73 | ||
| AK171458.1 | - | 1 - 2 | 169 | 73 | ||
| BC138694.1 | - | 1 - 2 | 143 | 8861716 | 73 | |
| BC138695.1 | - | 1 - 2 | 143 | 8861717 | 73 | |
| BC145384.1 | - | 1 - 2 | 143 | 73 | ||
Set Membership: |
|