Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03990600_m1
          See other PTX4 GE Assays ›
          Gene Symbol:
          Ptx4
          Gene Name:
          pentraxin 4
          Gene Aliases:
          1110018H23Rik
          Chromosome Location:
          Chr.17: 25120697 - 25125286 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03990600_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163416.1 NP_001156888.1 2 - 3 800 101
          NM_026747.1 NP_081023.1 2 - 3 890 101
          GenBank mRNA AK003777.1 - 1 - 2 359 101
          BC107380.2 - 2 - 3 783 40057084 101
          BC107381.2 - 2 - 3 873 40057088 101
          BC118508.1 - 2 - 3 873 40106397 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000361.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156800 BC118508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156800 BC107380.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156800 BC107381.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156800 BC115902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090909 AK003777.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090909 NM_001163416.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090909 NM_026747.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381638 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163589 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156800 BC118508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156800 BC107380.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156800 BC107381.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156800 BC115902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090909 AK003777.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090909 NM_001163416.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090909 NM_026747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123422 BC118508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123422 BC107380.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123422 BC107381.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123422 BC115902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123422 AK003777.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442687 - -
          Affymetrix HT MG-430 PM Array Plate 1430194_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442683 - -
          Mouse Genome 430 2.0 Array 1430194_PM_at - -
          Mouse Genome 430 2.0 Array 1430194_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68509_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68509_at - -
          Affymetrix Mouse Clariom S Array PSR1700002924.mm.1 NM_026747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002924.mm.1 NM_026747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002924.mm.1 TC1700000361.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002927.mm.1 NM_001163416.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002927.mm.1 NM_001163416.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002927.mm.1 NM_026747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002927.mm.1 NM_026747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002927.mm.1 TC1700000361.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002929.mm.1 NM_001163416.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002929.mm.1 NM_001163416.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002929.mm.1 NM_026747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002929.mm.1 NM_026747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002929.mm.1 TC1700000361.mm.1 MTA TC
          Mouse Expression Set 430 1430194_PM_at - -
          Mouse Expression Set 430 1430194_at - -
          Murine Genome U74v2 Set 171119_at - -
          Murine Genome U74v2 Set 171119_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430194_at - -
          Affymetrix HT Mouse Genome 430B Array 1430194_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 758024 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218798 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218798 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline