Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03990724_m1
          See other PRRG1 GE Assays ›
          Gene Symbol:
          Prrg1
          Gene Name:
          proline rich Gla (G-carboxyglutamic acid) 1
          Gene Aliases:
          2010007L08Rik
          Chromosome Location:
          Chr.X: 78449610 - 78583913 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03990724_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prrg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164275.2 NP_001157747.1 3 - 4 304 100
          NM_001289683.1 NP_001276612.1 4 - 5 400 100
          NM_027322.2 NP_081598.1 3 - 4 331 100
          GenBank mRNA AK135895.1 - 4 - 5 395 100
          CJ052035.1 - 3 - 4 329 100
          CX242928.1 - 3 - 4 282 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002513.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009172 NM_027322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009172 NM_001164275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P497188 NM_027322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P497188 NM_001164275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P497188 NM_027322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P497188 NM_001164275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004457 AK135895.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004457 NM_027322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004457 NM_001164275.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112669 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P497188 NM_027322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P497188 NM_001164275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P497188 NM_027322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P497188 NM_001164275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009172 NM_027322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009172 NM_001164275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004457 AK135895.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004457 NM_027322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004457 NM_001164275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P684989 AK083396.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183086 AK008151.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14594635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15083967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15172111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14697306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14516192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14602471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15083978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32481899 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605496 - -
          Affymetrix HT MG-430 PM Array Plate 1455724_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441441_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431484_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605493 - -
          Mouse Genome 430 2.0 Array 1431484_at - -
          Mouse Genome 430 2.0 Array 1455724_at - -
          Mouse Genome 430 2.0 Array 1455724_PM_at - -
          Mouse Genome 430 2.0 Array 1431484_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00016038.mm.1 NM_001164275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016038.mm.1 NM_001289683.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016038.mm.1 NM_027322.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016038.mm.1 TC0X00002513.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016047.mm.1 NM_001164275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016047.mm.1 NM_001289683.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016047.mm.1 NM_027322.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016047.mm.1 TC0X00002513.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016064.mm.1 NM_001164275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016064.mm.1 TC0X00002513.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016062.mm.1 NM_001289683.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016062.mm.1 TC0X00002513.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016037.mm.1 NM_001164275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016037.mm.1 NM_001289683.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016037.mm.1 NM_027322.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016037.mm.1 TC0X00002513.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016039.mm.1 NM_001164275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016039.mm.1 NM_001289683.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016039.mm.1 NM_027322.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016039.mm.1 TC0X00002513.mm.1 MTA TC
          Mouse Expression Set 430 1431484_at - -
          Mouse Expression Set 430 1431484_PM_at - -
          Mouse Expression Set 430 1455724_at - -
          Mouse Expression Set 430 1455724_PM_at - -
          Murine Genome U74v2 Set 168543_r_at - -
          Murine Genome U74v2 Set 168543_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1441441_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455724_at - -
          Affymetrix HT Mouse Genome 430B Array 1441441_at - -
          Affymetrix HT Mouse Genome 430B Array 1455724_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431484_at - -
          Affymetrix HT Mouse Genome 430B Array 1431484_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 830275 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100380044 - -
          Illumina mouse-6 v1.0 expression beadchip 100380044 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2521621 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214039 - -
          mouse-6 v1.1 (Illumina) scl0002993.1_1671-S - -
          Illumina Sentrix mouse-6.v1 bead array 100380044 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100380044 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236077 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline