| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900001210.mm.1 |
NM_001165919.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001210.mm.1 |
NM_001165919.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001210.mm.1 |
TC1900000105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001212.mm.1 |
NM_001165919.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001212.mm.1 |
NM_001165919.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001212.mm.1 |
TC1900000105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001211.mm.1 |
NM_001165919.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001211.mm.1 |
NM_001165919.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001211.mm.1 |
TC1900000105.mm.1 |
MTA TC |