Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03991048_m1
          See other SKINT11 GE Assays ›
          Gene Symbol:
          Skint11
          Gene Name:
          selection and upkeep of intraepithelial T cells 11
          Gene Aliases:
          A630098G03Rik, Gm569
          Chromosome Location:
          Chr.4: 114163384 - 114245028 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03991048_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Skint11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166027.1 NP_001159499.1 5 - 6 781 68
          GenBank mRNA EU099309.1 - 5 - 6 752 68

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 EU099309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 NM_001166027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 NM_177669.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 AK042513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 EU099309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 NM_001166027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 NM_177669.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P169595 AK042513.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128134 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124269 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134753 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 EU099309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 NM_001166027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 NM_177669.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 AK042513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 EU099309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 NM_001166027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 NM_177669.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P169595 AK042513.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23218142 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134753 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00963016 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05363850 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05363875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05363855 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124269 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05363898 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05363851 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05363904 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05363879 - -
          Whole Mouse Genome G4122A A_52_P169595 EU099309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169595 NM_001166027.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169595 NM_177669.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169595 AK042513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169595 EU099309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169595 NM_001166027.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169595 NM_177669.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169595 AK042513.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05363879 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134753 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05363843 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00963016 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03965734 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03965726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23218137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03829177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03297240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03685658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03245118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03297236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23218124 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10507096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507097 - -
          Affymetrix Mouse Gene 1.1 ST Array 10507095 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2763037 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2763037 - -
          mouse-6 v1.1 (Illumina) scl25108.1.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25108.1.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2763039 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2763039 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline