Search
Search

Cst12| Gene Symbol: Cst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027054.1 | NP_081330.1 | 1 - 2 | 228 | 85 | |
| GenBank mRNA | AF440737.2 | - | 1 - 2 | 210 | 85 | |
| AK005670.1 | - | 1 - 2 | 219 | 85 | ||
| BC119432.2 | - | 1 - 2 | 228 | 40041153 | 85 | |
Set Membership: |
|