Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03991515_s1
          See other H2BFM GE Assays ›
          Gene Symbol:
          H2bfm
          Gene Name:
          H2B histone family, member M
          Gene Aliases:
          1700014N06Rik, H2bfw, H2bl2
          Chromosome Location:
          Chr.X: 136927325 - 136928373 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03991515_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2bfm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027067.2 NP_081343.1 1 554 80
          GenBank mRNA AK005982.1 - 1 555 80
          BC117076.1 - 1 545 40126395 80
          BC117080.1 - 1 545 40126399 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683274 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683275 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958047 BC117080.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958047 BC117076.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192577 AK005982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192577 NM_027067.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500529 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142497 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2192577 AK005982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2192577 NM_027067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958047 BC117080.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958047 BC117076.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142497 - -
          Whole Mouse Genome G4122A A_51_P401594 BC117080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401594 AK005982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401594 BC117076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401594 NM_027067.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120526 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683274 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32683280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14634465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14634463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14997876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14634475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14634464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14807876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32683284 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601902 - -
          Affymetrix HT MG-430 PM Array Plate 1454199_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10601901 - -
          Mouse Genome 430 2.0 Array 1454199_PM_at - -
          Mouse Genome 430 2.0 Array 1454199_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69389_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69389_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00008460.mm.1 NM_027067.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008460.mm.1 NM_027067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008460.mm.1 TC0X00001341.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008459.mm.1 NM_027067.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008459.mm.1 NM_027067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008459.mm.1 TC0X00001341.mm.1 MTA TC
          Mouse Expression Set 430 1454199_at - -
          Mouse Expression Set 430 1454199_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454199_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454199_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 835701 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106590373 - -
          Illumina mouse-6 v1.0 expression beadchip 106590373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259771 - -
          Illumina Sentrix mouse-6.v1 bead array 106590373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106590373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259771 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0