Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03991747_s1
          See other HIST3H2BA GE Assays ›
          Gene Symbol:
          Hist3h2ba
          Gene Name:
          histone cluster 3, H2ba
          Gene Aliases:
          1500011O09Rik, AI413321
          Chromosome Location:
          Chr.11: 58948911 - 58949372 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03991747_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist3h2ba
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK018765.1 - 1 551 91
          BC051921.1 - 1 541 5684450 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          histone

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833690 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036813 NM_030082.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036813 AK018765.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036813 BC051921.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41742 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036813 NM_030082.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036813 AK018765.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036813 BC051921.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P338664 NM_030082.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P338664 AK018765.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P338664 BC051921.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833665 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07358569 - -
          Whole Mouse Genome G4122A A_51_P338664 NM_030082.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338664 AK018765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338664 BC051921.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09365490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09433808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09433817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09365492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09433818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09433816 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09433807 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09278532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09433808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09365494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09410601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09268085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09365492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09624419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09631792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09268093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09365490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376445 - -
          Affymetrix HT MG-430 PM Array Plate 1449482_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376444 - -
          Mouse Genome 430 2.0 Array 1449482_at - -
          Mouse Genome 430 2.0 Array 1449482_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78303_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78303_at - -
          Mouse Expression Set 430 1449482_at - -
          Mouse Expression Set 430 1449482_PM_at - -
          Mouse Genome 430A 2.0 Array 1449482_at - -
          Mouse Genome 430A 2.0 Array 1449482_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449482_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449482_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1990204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2652022 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2652022 - -
          mouse-6 v1.1 (Illumina) scl41509.1.1_222-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41509.1.1_222-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2652022 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2652022 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41509.1.1_222-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline