Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03991865_g1
          See other OLFR1274-PS GE Assays ›
          Gene Symbol:
          Olfr1274-ps
          Gene Name:
          olfactory receptor 1274, pseudogene
          Gene Aliases:
          GA_x5J8B7W2BV0-3116-4045, MOR228-4, MOR228-5, Olfr1274
          Chromosome Location:
          Chr.2: 90400640 - 90401674 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03991865_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1274-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146263.2 NP_666375.2 1 954 109
          GenBank mRNA BC145937.1 - 954 40131240 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_51_P283359 NM_146263.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283359 BC145937.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138413 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21416889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21416894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21416889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01880234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02073737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01372030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21416892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473646 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484875 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473648 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473643 - -
          Affymetrix Mouse Clariom S Array PSR0200010183.mm.1 NM_146263.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010183.mm.1 NM_146263.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010183.mm.1 TC0200001348.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 579802 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5360601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219722 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219722 - -
          mouse-6 v1.1 (Illumina) scl00257663.1_248-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00257663.1_248-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360601 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219722 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219722 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-djsrz:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline