Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03992707_s1
          See other GCNT4 GE Assays ›
          Gene Symbol:
          Gcnt4
          Gene Name:
          glucosaminyl (N-acetyl) transferase 4, core 2 (beta-1,6-N-acetylglucosaminyltransferase)
          Gene Aliases:
          C2gnt3, Gm279, Gm73
          Chromosome Location:
          Chr.13: 96924352 - 96950914 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03992707_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gcnt4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166065.1 NP_001159537.1 3 1650 106
          GenBank mRNA AK157788.1 - 3 1651 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          inter-male aggressive behavior
          thyroid hormone metabolic process
          tissue morphogenesis
          homeostasis of number of cells
          kidney morphogenesis
          beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity
          acetylglucosaminyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966967 NM_001166065.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966967 AK157788.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504017 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 180300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170953 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966967 NM_001166065.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966967 AK157788.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02752969 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02752969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10776966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11072120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10500903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11108705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10718290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10438916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11074947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10709727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10599063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10709737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11108710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10479605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10481942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11072121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29248579 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10406779 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406780 - -
          Affymetrix Mouse Gene 1.1 ST Array 10406777 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687739 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0