Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03992792_s1
          See other RPS7 GE Assays ›
          Gene Symbol:
          Rps7
          Gene Name:
          ribosomal protein S7
          Gene Aliases:
          Mtu, Rps7A, S7
          Chromosome Location:
          Chr.12: 28630847 - 28635953 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03992792_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011300.3 NP_035430.1 7 753 102
          GenBank mRNA AA221266.1 - 2 254 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps7

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          rRNA processing
          translation
          cell differentiation
          ribosomal small subunit biogenesis
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314236 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314235 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314234 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314237 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055411 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485864 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158978 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055411 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127545 BC100397.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127545 AA221266.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127545 AK135520.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127545 NM_011300.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127545 AK160632.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28314176 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07571204 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158978 - -
          Whole Mouse Genome G4122A A_51_P127545 BC100397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127545 AA221266.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127545 AK135520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127545 NM_011300.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127545 AK160632.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10297848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10058918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10039268 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10039265 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314235 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314236 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314237 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28314234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28314209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10089337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10384502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10039265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10009299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10410429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10009310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10058919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28314245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10399766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607392 - -
          Affymetrix HT MG-430 PM Array Plate 1419364_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1435593_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461389 - -
          Affymetrix Mouse Gene 1.1 ST Array 10399760 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607391 - -
          Mouse Genome 430 2.0 Array 1435593_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455364_PM_a_at - -
          Mouse Genome 430 2.0 Array 1455364_a_at - -
          Mouse Genome 430 2.0 Array 1435593_x_at - -
          Mouse Genome 430 2.0 Array 1419364_PM_a_at - -
          Mouse Genome 430 2.0 Array 1419364_a_at - -
          Affymetrix Mouse Clariom S Array PSR1200010867.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010867.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010867.mm.1 TC1200001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010866.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010866.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010866.mm.1 TC1200001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010864.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010864.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010864.mm.1 TC1200001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010868.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010868.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010868.mm.1 TC1200001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010865.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010865.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010865.mm.1 TC1200001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010869.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010869.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010869.mm.1 TC1200001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010863.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010863.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010863.mm.1 TC1200001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010873.mm.1 NM_011300.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010873.mm.1 NM_011300.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010873.mm.1 TC1200001568.mm.1 MTA TC
          Mouse Expression Set 430 1455364_a_at - -
          Mouse Expression Set 430 1419364_a_at - -
          Mouse Expression Set 430 1435593_PM_x_at - -
          Mouse Expression Set 430 1435593_x_at - -
          Mouse Expression Set 430 1455364_PM_a_at - -
          Mouse Expression Set 430 1419364_PM_a_at - -
          Murine Genome U74v2 Set 101212_PM_at - -
          Murine Genome U74v2 Set 101212_at - -
          Mouse Genome 430A 2.0 Array 1455364_a_at - -
          Mouse Genome 430A 2.0 Array 1419364_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1455364_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1419364_a_at - -
          Mouse Genome 430A 2.0 Array 1435593_x_at - -
          Mouse Genome 430A 2.0 Array 1435593_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1435593_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1435593_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6860113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860113 - -
          mouse-6 v1.1 (Illumina) scl020115.2_55-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020115.2_55-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2916116 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710068 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2916116 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020115.2_55-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0