Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04178117_s1
          See other OLFR1031 GE Assays ›
          Gene Symbol:
          Olfr1031
          Gene Name:
          olfactory receptor 1031
          Gene Aliases:
          MOR200-1
          Chromosome Location:
          Chr.2: 85991819 - 85992829 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04178117_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1031
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011759.2 NP_001011759.1 1 649 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1031

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140413 NM_001011759.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140413 NM_001011759.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400296 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04593965 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01700298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01700310 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01816815 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01700303 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01700309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01816814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01960844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01700308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01415664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01700319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01897193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01816813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01150922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01700298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01150934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02024120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01747262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400290 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473488 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473487 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 726368 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160975 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160975 - -
          mouse-6 v1.1 (Illumina) GI_58801305-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160975 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline