| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
291994 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
171288 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309896 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471503 |
AK010744.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471503 |
AK090346.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471503 |
AK153252.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471503 |
BC012711.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471503 |
D49744.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471503 |
NM_008033.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471503 |
XM_001477383.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
332980 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
94478 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
860125 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
546513 |
AK042514.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
182033 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
508349 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
125423 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
102020 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
30644 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P148835 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P163633 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P232859 |
AK137753.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P232859 |
AY137341.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P232859 |
AY137342.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P232859 |
BC117543.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P232859 |
NM_027562.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841358 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841330 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841346 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841357 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841327 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841331 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841352 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841328 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841348 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24841329 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06066973 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P148835 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01445044 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06066967 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06066940 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P163633 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P243418 |
NM_027562.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P243418 |
AK014570.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P243418 |
NM_001168223.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P243418 |
NM_001168224.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P232859 |
AK137753.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P232859 |
AY137341.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P232859 |
AY137342.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P232859 |
BC117543.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P232859 |
NM_027562.4 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117503 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06066973 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P163633 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P148835 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05249572 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05249571 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05249558 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05249561 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05249564 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05249568 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05249563 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841331 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841402 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841397 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841364 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841329 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841391 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841413 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841539 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841434 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841325 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841404 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841327 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841410 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841382 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841430 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841556 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841462 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841328 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841403 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841361 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841358 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841558 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841545 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841547 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841459 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841441 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841432 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841417 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841552 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841389 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841415 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841461 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841323 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841488 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841330 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841362 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841412 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841486 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841400 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841348 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841384 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841439 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841398 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24841531 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841448 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841350 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05058749 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841328 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841488 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841498 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05082024 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05818802 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841486 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841481 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05818803 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841441 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05718925 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841451 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841541 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841545 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841552 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841327 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05248915 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841357 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841491 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841444 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841558 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05284814 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841439 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841474 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841331 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841485 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841330 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841459 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841539 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05343522 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841435 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841360 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841482 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841347 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841346 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841457 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841434 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841462 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841496 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841538 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841556 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841348 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841432 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841447 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841358 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841461 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841560 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841352 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24841547 |
- |
- |