Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04179654_m1
          See other ATF5 GE Assays ›
          Gene Symbol:
          Atf5
          Gene Name:
          activating transcription factor 5
          Gene Aliases:
          AFTA, Atf7, Atfx, ODA-10
          Chromosome Location:
          Chr.7: 44812256 - 44816658 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04179654_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atf5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030693.2 NP_109618.1 2 - 3 471 69
          NR_033136.1 2 - 3 551 69
          GenBank mRNA AF310624.1 - 2 - 3 492 69
          AF312938.1 - 2 - 3 447 69
          AF375475.1 - 2 - 3 476 69
          AF375476.1 - 2 - 3 471 69
          AK131939.1 - 2 - 3 492 69
          AK146479.1 - 2 - 3 493 69
          AK159624.1 - 2 - 3 495 68
          BC010195.1 - 1 - 2 304 3157499 69
          BC092068.1 - 2 - 3 476 4210931 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002974.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atf5

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          post-embryonic development
          regulation of gene expression
          olfactory bulb interneuron differentiation
          olfactory bulb interneuron development
          olfactory lobe development
          multicellular organism growth
          regulation of cell proliferation
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of astrocyte differentiation
          negative regulation of neurogenesis
          regulation of cell cycle
          RNA polymerase II regulatory region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          heat shock protein binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089414 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105858 BC092068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105858 BC010195.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105858 AK131939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105858 AK159624.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105858 AK160085.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4700 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148940 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105858 BC092068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105858 BC010195.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105858 AK131939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105858 AK159624.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105858 AK160085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269216 BC092068.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269216 BC010195.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269216 AK131939.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269216 AK146479.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269216 AK159624.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25089429 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01506611 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148940 - -
          Whole Mouse Genome G4122A A_51_P269216 BC092068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269216 BC010195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269216 AK131939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269216 AK146479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269216 AK159624.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103428 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134190 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06060550 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06060552 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06315866 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06315867 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06318062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06454080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06646415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06489196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06361452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06289644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562910 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562909 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562908 - -
          Affymetrix HT MG-430 PM Array Plate 1425927_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562905 - -
          Mouse Genome 430 2.0 Array 1425927_PM_a_at - -
          Mouse Genome 430 2.0 Array 1425927_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107503_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107503_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700026510.mm.1 NM_030693.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026510.mm.1 NM_030693.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026510.mm.1 NR_033136.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026510.mm.1 NR_033136.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026510.mm.1 TC0700002974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026512.mm.1 NM_030693.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026512.mm.1 NM_030693.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026512.mm.1 TC0700002974.mm.1 MTA TC
          Mouse Expression Set 430 1425927_PM_a_at - -
          Mouse Expression Set 430 1425927_a_at - -
          Murine Genome U74v2 Set 103006_at - -
          Murine Genome U74v2 Set 103006_PM_at - -
          Mouse Genome 430A 2.0 Array 1425927_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1425927_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425927_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425927_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5670685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255341 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255341 - -
          mouse-6 v1.1 (Illumina) scl31408.4_131-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31408.4_131-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646985 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646985 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31408.4_131-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline