Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04179808_s1
          See other OLFR319 GE Assays ›
          Gene Symbol:
          Olfr319
          Gene Name:
          olfactory receptor 319
          Gene Aliases:
          MOR285-2
          Chromosome Location:
          Chr.11: 58701703 - 58702623 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04179808_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr319
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146500.2 NP_666711.2 1 547 137
          GenBank mRNA BC104265.1 - 1 547 40040340 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr319

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986665 BC104265.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855038 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152641 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986665 BC104265.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P441874 BC104265.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P441874 NM_146500.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832326 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152641 - -
          Whole Mouse Genome G4122A A_52_P441874 BC104265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P441874 NM_146500.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100577 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107839 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09648110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09059225 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09162887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09059233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09059211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09059222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09162880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09162892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09162884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09162887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09082934 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10386208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376407 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376406 - -
          Affymetrix Mouse Gene 1.1 ST Array 10386207 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106590671 - -
          MouseWG-6 v2 Expression BeadChip 5050020 - -
          Illumina mouse-6 v1.0 expression beadchip 5050020 - -
          Illumina mouse-6 v1.0 expression beadchip 106590671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2728212 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2728212 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2535145 - -
          mouse-6 v1.1 (Illumina) scl14111.1.1_40-S - -
          mouse-6 v1.1 (Illumina) GI_33238879-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050020 - -
          Illumina Sentrix mouse-6.v1 bead array 106590671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl14111.1.1_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050020 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106590671 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2728212 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2728212 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl14111.1.1_40-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline