Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04179829_g1
          See other OLFR313 GE Assays ›
          Gene Symbol:
          Olfr313
          Gene Name:
          olfactory receptor 313
          Gene Aliases:
          MOR222-2
          Chromosome Location:
          Chr.11: 58816928 - 58818001 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04179829_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr313
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146536.2 NP_666747.2 1 947 176
          GenBank mRNA BC117002.1 - 1 947 40126321 176
          BC120742.1 - 1 947 40129665 176

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr313

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832905 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164599 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100246 - -
          Whole Mouse Genome G4122A A_51_P159573 BC104273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159573 NM_001011819.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159573 BC104268.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102682 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134456 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121205 BC104273.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121205 NM_001011819.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121205 BC104268.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164599 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100246 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09698567 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09446975 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09698565 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09446973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09361671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09108510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09051968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09363192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09363194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832946 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376415 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376418 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376416 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376414 - -
          Affymetrix Mouse Clariom S Array PSR1100005609.mm.1 NM_146536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005609.mm.1 NM_146536.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005609.mm.1 TC1100000703.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 721885 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5050100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775191 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775191 - -
          mouse-6 v1.1 (Illumina) scl41514.1.1_155-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41514.1.1_155-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2779272 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775191 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2779272 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41514.1.1_155-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline