Search
Search

Olfr1020| Gene Symbol: Olfr1020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146580.2 | NP_666791.1 | 1 | 655 | 78 | |
| GenBank mRNA | BC119145.1 | - | 1 | 653 | 8733894 | 78 |
| BC119147.1 | - | 1 | 655 | 8733896 | 78 | |
Set Membership: |
|