Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04180041_s1
          See other OLFR1221 GE Assays ›
          Gene Symbol:
          Olfr1221
          Gene Name:
          olfactory receptor 1221
          Gene Aliases:
          MOR233-3
          Chromosome Location:
          Chr.2: 89111575 - 89112510 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04180041_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1221
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146902.2 NP_667113.2 1 446 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1221

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358700 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166272 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358700 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358700 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166272 - -
          Whole Mouse Genome G4122A A_51_P358700 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122004 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02171274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412600 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484790 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484789 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 570903 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105270593 - -
          Illumina mouse-6 v1.0 expression beadchip 3520050 - -
          Illumina mouse-6 v1.0 expression beadchip 105270593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222921 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222921 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222072 - -
          mouse-6 v1.1 (Illumina) scl4292.1.1_191-S - -
          mouse-6 v1.1 (Illumina) GI_22129082-S - -
          Illumina Sentrix mouse-6.v1 bead array 105270593 - -
          Illumina Sentrix mouse-6.v1 bead array 3520050 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4292.1.1_191-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105270593 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520050 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222921 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222921 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline