Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04180506_g1
          See other D6ERTD527E GE Assays ›
          Gene Symbol:
          D6Ertd527e
          Gene Name:
          DNA segment, Chr 6, ERATO Doi 527, expressed
          Gene Aliases:
          AU043841
          Chromosome Location:
          Chr.6: 87100465 - 87113003 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04180506_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D6Ertd527e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167937.1 NP_001161409.1 2 656 97
          NM_001167938.1 NP_001161410.1 2 653 97
          GenBank mRNA AK139701.1 - 2 655 97
          AK139822.1 - 2 654 97
          BC157933.1 - 2 9088107 97

          Back To Top

          More Information


          Additional Information:

          This assay targets the same location as MTA transcript cluster(s) TC0600001003.mm
          This assay targets the same location as RTA transcript cluster(s) TC0400001330.rn

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 BC157933.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 NM_001167938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 AK139701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 AK139822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 NM_001167937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 BC157933.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 NM_001167938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 AK139701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 AK139822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P78931 NM_001167937.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812309 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420406 NM_147071.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815409 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 BC157933.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 NM_001167938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 AK139701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 AK139822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 NM_001167937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 BC157933.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 NM_001167938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 AK139701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 AK139822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P78931 NM_001167937.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05970677 - -
          Whole Mouse Genome G4122A A_52_P78931 BC157933.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 NM_001167938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 AK139701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 AK139822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 NM_001167937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 BC157933.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 NM_001167938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 AK139701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 AK139822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P78931 NM_001167937.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P05358349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05019745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05696750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05748447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05019739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05762089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05534861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05792773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05019732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05802089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24613896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05762087 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433484_PM_at - -
          Mouse Genome 430 2.0 Array 1433484_at - -
          Mouse Genome 430 2.0 Array 1433484_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52372_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52372_at - -
          Affymetrix Mouse Clariom S Array PSR0600007881.mm.1 NM_001167937.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007881.mm.1 NM_001167937.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007881.mm.1 NM_001167938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007881.mm.1 NM_001167938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007881.mm.1 TC0600001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007882.mm.1 NM_001167937.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007882.mm.1 NM_001167937.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007882.mm.1 NM_001167938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007882.mm.1 NM_001167938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007882.mm.1 TC0600001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007877.mm.1 NM_001167937.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007877.mm.1 NM_001167937.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007877.mm.1 NM_001167938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007877.mm.1 NM_001167938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007877.mm.1 TC0600001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007880.mm.1 NM_001167937.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007880.mm.1 NM_001167937.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007880.mm.1 NM_001167938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007880.mm.1 NM_001167938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007880.mm.1 TC0600001003.mm.1 MTA TC
          Mouse Expression Set 430 1433484_at - -
          Mouse Expression Set 430 1433484_PM_at - -
          Murine Genome U74v2 Set 131391_at - -
          Murine Genome U74v2 Set 131391_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433484_at - -
          Affymetrix HT Mouse Genome 430B Array 1433484_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline