| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800019655.mm.1 |
NM_001007571.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019655.mm.1 |
NM_001007571.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019655.mm.1 |
TC0800002578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019654.mm.1 |
NM_001007571.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019654.mm.1 |
NM_001007571.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019654.mm.1 |
TC0800002578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019651.mm.1 |
NM_001007571.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019651.mm.1 |
NM_001007571.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019651.mm.1 |
TC0800002578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019653.mm.1 |
NM_001007571.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019653.mm.1 |
NM_001007571.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019653.mm.1 |
TC0800002578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019656.mm.1 |
NM_001007571.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019656.mm.1 |
NM_001007571.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019656.mm.1 |
TC0800002578.mm.1 |
MTA TC |