Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04204063_s1
          See other OLFR605 GE Assays ›
          Gene Symbol:
          Olfr605
          Gene Name:
          olfactory receptor 605
          Gene Aliases:
          GA_x5J8B7TSRWK-430-5, MOR202-22P, MOR24-5
          Chromosome Location:
          Chr.7: 103442140 - 103443150 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04204063_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr605
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011854.2 NP_001011854.2 1 748 100
          GenBank mRNA BC147216.1 - 1 748 9056257 100
          BC147217.1 - 1 748 9056245 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr605

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135632 NM_001011854.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135632 BC147216.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135632 BC147217.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135632 NM_001011854.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135632 BC147216.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135632 BC147217.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300401 NM_001011854.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383349 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06114571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06434692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06161431 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06434687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06397035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06542134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383279 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566214 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566213 - -
          Affymetrix Mouse Clariom S Array PSR0700032708.mm.1 NM_001011854.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032708.mm.1 NM_001011854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032708.mm.1 TC0700003890.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 770030 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160282 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160282 - -
          mouse-6 v1.1 (Illumina) GI_58801465-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160282 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160282 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline