| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400030395.mm.1 |
TC0400003708.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030395.mm.1 |
NM_001017968.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030395.mm.1 |
NM_001017968.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030396.mm.1 |
TC0400003708.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030396.mm.1 |
NM_001017968.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030396.mm.1 |
NM_001017968.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030397.mm.1 |
TC0400003708.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030397.mm.1 |
NM_001017968.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030397.mm.1 |
NM_001017968.2 |
Refseq |