Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04204476_m1
          See other CEACAM1 GE Assays ›
          Gene Symbol:
          Ceacam1
          Gene Name:
          carcinoembryonic antigen-related cell adhesion molecule 1
          Gene Aliases:
          Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1, bb-1, mCEA1, mmCGM1, mmCGM1a, mmCGM2
          Chromosome Location:
          Chr.7: 25461702 - 25477625 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04204476_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ceacam1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039185.1 NP_001034274.1 5 - 6 1332 97
          NM_001039186.1 NP_001034275.1 5 - 6 1332 97
          NM_001039187.1 NP_001034276.1 3 - 4 792 97
          NM_011926.2 NP_036056.2 3 - 4 792 97
          GenBank mRNA AB236328.2 - 3 - 4 701 97
          AB236329.1 - 3 - 4 701 97
          AB236330.2 - 4 - 5 986 97
          BC016891.1 - 5 - 6 1284 97
          BC114998.1 - 1 - 2 264 97
          BC121189.1 - 4 - 5 889 97
          M77196.1 - 5 - 6 1310 97
          M96935.1 - 4 - 5 694 97
          X15351.1 - 5 - 6 1344 97
          X67278.1 - 3 - 4 804 97
          X67279.1 - 5 - 6 1344 97
          X67282.1 - 3 - 4 804 97

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ceacam1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine production
          homophilic cell adhesion via plasma membrane adhesion molecules
          viral process
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          negative regulation of interleukin-2 production
          positive regulation of T cell proliferation
          negative regulation of T cell proliferation
          positive regulation of MAP kinase activity
          negative regulation by host of viral process
          positive regulation by host of viral process
          negative regulation by host of viral genome replication
          negative regulation of JNK cascade
          positive regulation of JNK cascade
          viral entry into host cell
          Peyer's patch development
          positive regulation of immunoglobulin secretion
          positive regulation of NFAT protein import into nucleus
          positive regulation of activation-induced cell death of T cells
          positive regulation of CD4-positive, alpha-beta T cell activation
          positive regulation of CD4-positive, alpha-beta T cell proliferation
          positive regulation of CD8-positive, alpha-beta T cell activation
          virus receptor activity
          protein binding
          protein kinase binding
          protein homodimerization activity
          virion binding

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 520843 - -
          ABI Mouse Genome Survey Microarray 834228 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline