Search
Search
| Gene Symbol: Crnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081200.1 | NP_001074669.1 | 2 - 3 | 203 | 77 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BB618328.1 | - | 203 | Chr. - - [-] on Build | 77 | ||
Set Membership: |
|