Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04205030_gH
          See other GM11596 GE Assays ›
          Gene Symbol:
          Gm11596
          Gene Name:
          predicted gene 11596
          Gene Aliases:
          OTTMUSG00000002547
          Chromosome Location:
          Chr.11: 99792675 - 99793292 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04205030_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm11596
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099311.2 NP_001092781.1 1 148 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09626041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064407 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10390948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390935 - -
          Affymetrix Mouse Gene 1.1 ST Array 10390934 - -
          Affymetrix Mouse Clariom S Array PSR1100034342.mm.1 NM_001099311.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034342.mm.1 NM_001099311.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034342.mm.1 TC1100003737.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 751447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline