Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04205237_s1
          See other GM11567 GE Assays ›
          Gene Symbol:
          Gm11567
          Gene Name:
          predicted gene 11567
          Gene Aliases:
          OTTMUSG00000002206
          Chromosome Location:
          Chr.11: 99879187 - 99880229 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04205237_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm11567
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101613.1 NP_001095083.1 1 551 108
          GenBank mRNA AK003994.1 - 1 552 108

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992408 AK003994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992408 NM_001101613.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85746 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104421 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992408 AK003994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992408 NM_001101613.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104421 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28064818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09045942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09313731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09045930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09097175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09097181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28064832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381104 - -
          Affymetrix HT MG-430 PM Array Plate 1430509_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381105 - -
          Affymetrix Mouse Gene 1.1 ST Array 10381103 - -
          Mouse Genome 430 2.0 Array 1430509_at - -
          Mouse Genome 430 2.0 Array 1430509_PM_at - -
          Mouse Expression Set 430 1430509_at - -
          Mouse Expression Set 430 1430509_PM_at - -
          Mouse Expression Set 430 1422209_s_at - -
          Mouse Expression Set 430 1422209_PM_s_at - -
          Murine Genome U74v2 Set 170907_PM_at - -
          Murine Genome U74v2 Set 170907_at - -
          Mouse Genome 430A 2.0 Array 1430509_at - -
          Mouse Genome 430A 2.0 Array 1430509_PM_at - -
          Mouse Genome 430A 2.0 Array 1422209_s_at - -
          Mouse Genome 430A 2.0 Array 1422209_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1430509_at - -
          Affymetrix HT Mouse Genome 430A Array 1430509_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 390923 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0