| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700001756.mm.1 |
NM_001104542.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001756.mm.1 |
NM_001104542.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001756.mm.1 |
TC1700000220.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001755.mm.1 |
NM_001104542.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001755.mm.1 |
NM_001104542.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001755.mm.1 |
TC1700000220.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001759.mm.1 |
NM_001104542.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001759.mm.1 |
NM_001104542.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001759.mm.1 |
TC1700000220.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001757.mm.1 |
NM_001104542.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001757.mm.1 |
NM_001104542.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001757.mm.1 |
TC1700000220.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001758.mm.1 |
NM_001104542.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001758.mm.1 |
NM_001104542.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001758.mm.1 |
TC1700000220.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001754.mm.1 |
NM_001104542.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001754.mm.1 |
NM_001104542.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001754.mm.1 |
TC1700000220.mm.1 |
MTA TC |