| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500026223.mm.1 |
TC0500002962.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026223.mm.1 |
NM_001104622.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026223.mm.1 |
NM_001104622.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026224.mm.1 |
TC0500002962.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026224.mm.1 |
NM_001104622.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026224.mm.1 |
NM_001104622.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026225.mm.1 |
TC0500002962.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026225.mm.1 |
NM_001104622.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026225.mm.1 |
NM_001104622.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026222.mm.1 |
TC0500002962.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026222.mm.1 |
NM_001104622.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026222.mm.1 |
NM_001104622.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026221.mm.1 |
TC0500002962.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026221.mm.1 |
NM_001104622.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026221.mm.1 |
NM_001104622.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026220.mm.1 |
TC0500002962.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026220.mm.1 |
NM_001104622.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026220.mm.1 |
NM_001104622.1 |
Refseq |