Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206125_g1
          See other OLFR1335 GE Assays ›
          Gene Symbol:
          Olfr1335
          Gene Name:
          olfactory receptor 1335
          Gene Aliases:
          MOR259-12, MOR259-5, Olfr1334-ps1, Olfr1335-ps1
          Chromosome Location:
          Chr.4: 118808855 - 118809862 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04206125_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1335
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207703.1 NP_997586.1 1 853 98
          GenBank mRNA BC147439.1 - 1 935 9056081 98
          BC147453.1 - 1 935 9056095 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1335

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033150 NM_207703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033150 BC147439.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033150 BC147453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033151 NM_207703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033151 BC147439.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033151 BC147453.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518578 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033151 NM_207703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033151 BC147439.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033151 BC147453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033150 NM_207703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033150 BC147439.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033150 BC147453.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23242322 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03798838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03798839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23242329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03584124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04089554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03584126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04079532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03523730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03798838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23242314 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515829 - -
          Affymetrix Mouse Gene 1.1 ST Array 10515828 - -
          Affymetrix Mouse Clariom S Array PSR0400027600.mm.1 TC0400003404.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027600.mm.1 NM_207703.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027600.mm.1 NM_207703.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027599.mm.1 TC0400003404.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027599.mm.1 NM_207703.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027599.mm.1 NM_207703.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027598.mm.1 TC0400003404.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027601.mm.1 TC0400003404.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163087 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3163087 - -
          mouse-6 v1.1 (Illumina) GI_70794769-A - -
          MouseRef-8 v2 Expression BeadChip ILMN_3133658 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3163087 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3056687 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163087 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3056687 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3133658 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_70794769-A - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline