| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500005000.mm.1 |
TC0500000539.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500005000.mm.1 |
NM_001145433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500005000.mm.1 |
NM_001145433.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500005007.mm.1 |
TC0500000539.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500005007.mm.1 |
NM_001145433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500005007.mm.1 |
NM_001145433.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004999.mm.1 |
TC0500000539.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004999.mm.1 |
NM_001145433.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004999.mm.1 |
NM_001145433.1 |
Refseq |