| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300013916.mm.1 |
NM_001162911.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013916.mm.1 |
NM_001162911.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013916.mm.1 |
NM_175252.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013916.mm.1 |
NM_175252.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013916.mm.1 |
TC1300002768.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013914.mm.1 |
NM_001162911.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013914.mm.1 |
NM_001162911.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013914.mm.1 |
NM_175252.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013914.mm.1 |
NM_175252.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013914.mm.1 |
TC1300002768.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013917.mm.1 |
NM_175252.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013917.mm.1 |
NM_175252.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013917.mm.1 |
TC1300002768.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013915.mm.1 |
NM_001162911.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013915.mm.1 |
NM_001162911.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013915.mm.1 |
NM_175252.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013915.mm.1 |
NM_175252.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013915.mm.1 |
TC1300002768.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013919.mm.1 |
NM_001162911.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013919.mm.1 |
NM_001162911.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013919.mm.1 |
NM_175252.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013919.mm.1 |
NM_175252.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013919.mm.1 |
TC1300002768.mm.1 |
MTA TC |