Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206759_mH
          See other PYDC3 GE Assays ›
          Gene Symbol:
          Pydc3
          Gene Name:
          pyrin domain containing 3
          Gene Aliases:
          9830148g24rik, E430029J22Rik
          Chromosome Location:
          Chr.1: 173673628 - 173698392 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04206759_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pydc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162938.1 NP_001156410.1 5 - 6 1697 94
          GenBank mRNA AK036671.1 - 5 - 6 1467 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001634.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          double-stranded DNA binding
          transcription factor binding
          identical protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181222 NM_001162938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181222 AK036671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036627 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001233 AK145253.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001233 AK165868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128761 NM_001162938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128761 AK036671.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97135 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001233 AK145253.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001233 AK165868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036627 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128761 NM_001162938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128761 AK036671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181222 NM_001162938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181222 AK036671.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00440606 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351881 - -
          Affymetrix Mouse Gene 1.1 ST Array 10351880 - -
          Affymetrix Mouse Clariom S Array PSR0100013605.mm.1 NM_001162938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013605.mm.1 NM_001162938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013605.mm.1 TC0100001634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013604.mm.1 NM_001162938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013604.mm.1 NM_001162938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013604.mm.1 TC0100001634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013606.mm.1 NM_001162938.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013606.mm.1 NM_001162938.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013606.mm.1 TC0100001634.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105910520 - -
          Illumina mouse-6 v1.0 expression beadchip 102940332 - -
          Illumina mouse-6 v1.0 expression beadchip 105910520 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219896 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2568505 - -
          Illumina Sentrix mouse-6.v1 bead array 105910520 - -
          Illumina Sentrix mouse-6.v1 bead array 102940332 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105910520 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102940332 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2568505 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline