Search
Search
| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008304.2 | NP_032330.1 | 3 - 4 | 833 | 113 | |
| GenBank mRNA | AK011042.1 | - | 3 - 4 | 682 | 113 | |
| BC003929.1 | - | 2 - 3 | 231 | 113 | ||
| BC047144.1 | - | 3 - 4 | 699 | 6395969 | 113 | |
| U00674.1 | - | 3 - 4 | 833 | 113 | ||
Set Membership: |
|