Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207885_m1
          See other CSN2 GE Assays ›
          Gene Symbol:
          Csn2
          Gene Name:
          casein beta
          Gene Aliases:
          Csnb
          Chromosome Location:
          Chr.5: 87692619 - 87699429 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04207885_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286020.1 NP_001272949.1 7 - 8 743 84
          NM_001286021.1 NP_001272950.1 7 - 8 758 84
          NM_001286022.1 NP_001272951.1 6 - 7 719 84
          NM_001286023.1 NP_001272952.1 5 - 6 701 84
          NM_001286024.1 NP_001272953.1 5 - 6 698 84
          NM_009972.2 NP_034102.1 7 - 8 746 84
          GenBank mRNA AK052803.1 - 7 - 8 743 84
          AK052805.1 - 7 - 8 743 84
          AK052822.1 - 7 - 8 740 84
          AK085678.1 - 7 - 8 740 84
          AK085694.1 - 7 - 8 743 84
          AK085698.1 - 7 - 8 740 84
          AK085729.1 - 7 - 8 743 84
          AK085756.1 - 7 - 8 746 84
          AK142616.1 - 7 - 8 743 84
          AK142620.1 - 7 - 8 743 84
          AK142631.1 - 7 - 8 743 84
          AK142635.1 - 7 - 8 743 84
          AK164747.1 - 7 - 8 743 84
          AK164761.1 - 7 - 8 743 84
          AK164791.1 - 7 - 8 743 84
          AK164792.1 - 7 - 8 740 84
          AK164793.1 - 7 - 8 740 84
          AK164794.1 - 7 - 8 743 84
          BC003863.1 - 2 - 3 568 84
          BC003976.1 - 3 - 4 592 84
          BC013332.1 - 7 - 8 743 3670912 84
          BC019114.1 - 7 - 8 724 3483918 84
          BC019189.1 - 6 - 7 736 5028194 84
          BC020356.1 - 7 - 8 723 84
          BC020383.1 - 7 - 8 723 84
          BC021153.1 - 7 - 8 741 3670372 84
          BC050270.1 - 7 - 8 723 84
          BC057671.1 - 4 - 5 2535 84
          BC080709.1 - 7 - 8 727 3671502 84
          BC092098.1 - 8 - 9 631 84
          X04490.1 - 7 - 8 742 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002702.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csn2

          Panther Classification:

          Molecular Function -

          storage protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          calcium ion transport
          lactation
          negative regulation of catalytic activity
          negative regulation of lactation
          enzyme inhibitor activity
          transporter activity
          calcium ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500023863.mm.1 TC0500002702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023863.mm.1 NM_001286020.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023863.mm.1 NM_001286021.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023863.mm.1 NM_001286022.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023863.mm.1 NM_001286023.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023863.mm.1 NM_001286024.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023863.mm.1 NM_009972.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline